2-Chlor-4-cyanbenzolsulfonylchlorid

Lieferant: Thermo Scientific

Synonyme: 2-Chlor-4-cyanbenzolsulfochlorid, 2-Chlor-4-cyanbenzolsulfonsäurechlorid, 2-Chlor-4-cyanbenzol-1-sulfonylchlorid

SEW01377.0001 SEW01377.250
MAYBSEW01377.0001EA 0 EUR
MAYBSEW01377.0001 MAYBSEW01377.250
2-Chlor-4-cyanbenzolsulfonylchlorid
2-Chlor-4-cyanbenzolsulfonylchlorid
Formel: C₇H₃Cl₂NO₂S
Molecular Weight: 236,08 g/mol
MDL: MFCD00177089
CAS-Nummer: 254749-11-6
UN: 2923
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR