2-Chlorisonicotinonitril
Lieferant: COMBI-BLOCKS
Synonyme:
2-Chlor-4-cyanpyridin, 2-Chlor-4-pyridincarbonitril, 2-Chlor-4-pyridincarbonsäurenitril
BK-4524-100G
BK-4524-25G
COBBBK-4524-100GEA
170
EUR
COBBBK-4524-100G
COBBBK-4524-25G
2-Chlorisonicotinonitril
2-Chlorisonicotinonitril
Formel:
C₆H₃ClN₂ Molecular Weight: 138.56 g/mol |
MDL:
MFCD00174318 CAS-Nummer: 33252-30-1 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...