Methyl-2-chlor-4-fluorbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chlor-4-fluorbenzoesäuremethylester

CA-4363-25G CA-4363-5G CA-4363-100G
COBBCA-4363-25GEA 102 EUR
COBBCA-4363-25G COBBCA-4363-5G COBBCA-4363-100G
Methyl-2-chlor-4-fluorbenzoat
Methyl-2-chlor-4-fluorbenzoat
Formel: C₈H₆ClFO₂
Molecular Weight: 188,59 g/mol
MDL: MFCD00173937
CAS-Nummer: 85953-29-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR