Methyl-2-chlor-4-fluorbenzoat

Lieferant: FLUOROCHEM

Synonyme: 2-Chlor-4-fluorbenzoesäuremethylester

006786-1G 006786-5G 006786-25G
FLUO006786-1GEA 30.6 EUR
FLUO006786-1G FLUO006786-5G FLUO006786-25G
Methyl-2-chlor-4-fluorbenzoat
Methyl-2-chlor-4-fluorbenzoat
Formel: C₈H₆ClFO₂
Molecular Weight: 188,59 g/mol
MDL: MFCD00173937
CAS-Nummer: 85953-29-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR