2-Chlor-4-fluorphenylboronsäure
Lieferant: COMBI-BLOCKS
BB-2193-5G
BB-2193-25G
BB-2193-1G
COBBBB-2193-5GEA
63.4
EUR
COBBBB-2193-5G
COBBBB-2193-25G
COBBBB-2193-1G
2-Chlor-4-fluorphenylboronsäure
2-Chlor-4-fluorphenylboronsäure
Formel:
C₆H₅BClFO₂ Molecular Weight: 174,37 g/mol |
MDL:
MFCD02684295 CAS-Nummer: 313545-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...