2-Chlor-4-methoxypyridin
Lieferant: COMBI-BLOCKS
PY-7265-5G
PY-7265-1G
COBBPY-7265-5GEA
0
EUR
COBBPY-7265-5G
COBBPY-7265-1G
2-Chlor-4-methoxypyridin
2-Chloro-4-methoxypyridine
Formel:
C₆H₆ClNO Siedepunkt: 229…230 °C (1013 hPa) Dichte: 1,258 g/cm³ (20 °C) |
MDL:
MFCD02093951 CAS-Nummer: 17228-69-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...