2-Chlorisonicotinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chlorpyridin-4-carbonsäure, 2-Chlor-4-pyridincarbonsäure

CA-4039-25G CA-4039-100G
COBBCA-4039-25GEA 0 EUR
COBBCA-4039-25G COBBCA-4039-100G
2-Chlorisonicotinsäure
2-Chlorisonicotinsäure
Formel: C₆H₄ClNO₂
MDL: MFCD00191402
CAS-Nummer: 6313-54-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR