2-Chlorisonicotinamid
Lieferant: COMBI-BLOCKS
PY-7835-1G
PY-7835-25G
PY-7835-5G
COBBPY-7835-1GEA
38.2
EUR
COBBPY-7835-1G
COBBPY-7835-25G
COBBPY-7835-5G
2-Chlorisonicotinamid
2-Chlorisonicotinamid
Formel:
C₆H₅ClN₂O Molecular Weight: 156.57 g/mol |
MDL:
MFCD00221401 CAS-Nummer: 100859-84-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...