2-Chlor-5-(trifluormethyl)benzolsulfonylchlorid

Lieferant: Thermo Scientific

Synonyme: 2-Chlor-5-(trifluormethyl)benzol-1-sulfonylchlorid, 2-Chlor-5-(trifluormethyl)benzolsulfochlorid, 2-Chlor-5-(trifluormethyl)benzolsulfonsäurechlorid

TL00339DA TL00339EA
MAYBTL00339DAEA 0 EUR
MAYBTL00339DA MAYBTL00339EA
2-Chlor-5-(trifluormethyl)benzolsulfonylchlorid
2-Chlor-5-(trifluormethyl)benzolsulfonylchlorid
Formel: C₇H₃Cl₂F₃O₂S
Molecular Weight: 279,07 g/mol
MDL: MFCD00052310
CAS-Nummer: 54090-08-3
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR