6-Chlornicotinonitril

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chlor-5-pyridincarbonitril, 6-Chlornicotinsäurenitril, 2-Chlor-5-cyanpyridin, 2-Chlor-5-pyridincarbonsäurenitril

CA-4304-25G CA-4304-100G CA-4304-5G
COBBCA-4304-25GEA 121 EUR
COBBCA-4304-25G COBBCA-4304-100G COBBCA-4304-5G
6-Chlornicotinonitril
6-Chlornicotinonitril
Formel: C₆H₃ClN₂
Molecular Weight: 138.56 g/mol
MDL: MFCD00084941
CAS-Nummer: 33252-28-7
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR