2-Chlor-5-fluorbenzoesäure

Lieferant: COMBI-BLOCKS
Achtung

CA-4098-25G CA-4098-100G CA-4098-5G
COBBCA-4098-25GEA 93.7 EUR
COBBCA-4098-25G COBBCA-4098-100G COBBCA-4098-5G
2-Chlor-5-fluorbenzoesäure
2-Chloro-5-fluorobenzoic acid
Formel: C₇H₄ClFO₂
Molecular Weight: 174,56 g/mol
MDL: MFCD00156967
CAS-Nummer: 2252-50-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR