2-Chlor-5-hydroxypyridin

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 2-Chloro-5-pyridinol, 6-Chloro-3-pyridinol

PY-7270-5G PY-7270-25G PY-7270-1G
COBBPY-7270-5GEA 50.9 EUR
COBBPY-7270-5G COBBPY-7270-25G COBBPY-7270-1G
2-Chlor-5-hydroxypyridin
2-Chlor-5-hydroxypyridin
Formel: C₅H₄ClNO
Molecular Weight: 129,55 g/mol
MDL: MFCD00234050
CAS-Nummer: 41288-96-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR