2-Chlor-5-methyl-3-nitropyridin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chlor-3-nitro-5-picolin

OT-0059-100G OT-0059-5G OT-0059-25G OT-0059-1G
COBBOT-0059-100GEA 310 EUR
COBBOT-0059-100G COBBOT-0059-5G COBBOT-0059-25G COBBOT-0059-1G
2-Chlor-5-methyl-3-nitropyridin
2-Chloro-5-methyl-3-nitropyridine
Formel: C₆H₅ClN₂O₂
Molecular Weight: 172,57 g/mol
MDL: MFCD02070020
CAS-Nummer: 23056-40-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR