2-Chlor-5-methyl-3-nitropyridin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Chlor-3-nitro-5-picolin
M-Clarity™ Qualitätsstufe = MQ100
705918-1G
705918-5G
SIAL705918-1GEA
25
EUR
SIAL705918-1G
SIAL705918-5G
2-Chlor-5-methyl-3-nitropyridin, Sigma-Aldrich®
2-Chloro-5-methyl-3-nitropyridine
Formel:
C₆H₅ClN₂O₂ Molecular Weight: 172,57 g/mol |
MDL:
MFCD02070020 CAS-Nummer: 23056-40-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...