2-Chlor-5-methylpyridin
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
2-Chloro-5-picoline
OS-7529-100G
OS-7529-25G
OS-7529-5G
COBBOS-7529-100GEA
130
EUR
COBBOS-7529-100G
COBBOS-7529-25G
COBBOS-7529-5G
2-Chlor-5-methylpyridin
2-Chlor-5-methylpyridin
Formel:
C₆H₆ClN Molecular Weight: 127.57 g/mol Siedepunkt: 97 °C (30 mmHg) Dichte: 1.169 g/cm³ (20 °C) |
MDL:
MFCD00792460 CAS-Nummer: 18368-64-4 EINECS: 418-050-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...