2-Chlor-5-nitrobenzaldehyd

Lieferant: COMBI-BLOCKS

OS-7614-5G OS-7614-25G OS-7614-100G
COBBOS-7614-5GEA 25.5 EUR
COBBOS-7614-5G COBBOS-7614-25G COBBOS-7614-100G
2-Chlor-5-nitrobenzaldehyd
2-Chloro-5-nitrobenzaldehyde
Formel: C₇H₄ClNO₃
Molecular Weight: 185.57 g/mol
Schmelzpunkt: 75…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007293
CAS-Nummer: 6361-21-3
EINECS: 228-830-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR