2-Chlor-5-nitropyridin

Lieferant: COMBI-BLOCKS
Achtung

OT-0346-25G OT-0346-100G
COBBOT-0346-25GEA 51.3 EUR
COBBOT-0346-25G COBBOT-0346-100G
2-Chlor-5-nitropyridin
2-Chloro-5-nitropyridine
Formel: C₅H₃ClN₂O₂
Molecular Weight: 158,54 g/mol
Schmelzpunkt: 107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006240
CAS-Nummer: 4548-45-2
EINECS: 224-908-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR