2-Chlor-6-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chlor-6-fluorbenzolboronsäure, Dihydroxy(2-chlor-6-fluorphenyl)boran, (2-Chlor-6-fluorphenyl)metaborsäure

BB-2387-5G BB-2387-25G BB-2387-1G
COBBBB-2387-5GEA 76.1 EUR
COBBBB-2387-5G COBBBB-2387-25G COBBBB-2387-1G
2-Chlor-6-fluorphenylboronsäure
2-Chlor-6-fluorphenylboronsäure
Formel: C₆H₅BClFO₂
Molecular Weight: 174,37 g/mol
MDL: MFCD04039892
CAS-Nummer: 313545-32-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR