2-Chlor-6-fluorpyridin

Lieferant: COMBI-BLOCKS

OS-7524-5G OS-7524-25G OS-7524-100G
COBBOS-7524-5GEA 21.9 EUR
COBBOS-7524-5G COBBOS-7524-25G COBBOS-7524-100G
2-Chlor-6-fluorpyridin
2-Chloro-6-fluoropyridine
Formel: C₅H₃ClFN
Molecular Weight: 131.54 g/mol
MDL: MFCD00077483
CAS-Nummer: 20885-12-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR