2-Chlor-6-methoxy-3-nitropyridin

Lieferant: COMBI-BLOCKS
Achtung

PY-7359-25G PY-7359-1G PY-7359-5G
COBBPY-7359-25GEA 63.4 EUR
COBBPY-7359-25G COBBPY-7359-1G COBBPY-7359-5G
2-Chlor-6-methoxy-3-nitropyridin
2-Chloro-6-methoxy-3-nitropyridine
Formel: C₆H₅ClN₂O₃
Molecular Weight: 188.57 g/mol
MDL: MFCD00130268
CAS-Nummer: 38533-61-8
EINECS: 253-989-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR