2-Chlor-6-methyl-3-nitropyridin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
6-Chlor-5-nitro-2-picolin, 6-Chlor-2-methyl-5-nitropyridin, 2-Chlor-3-nitro-6-picolin
360830010
360830050
ACRO360830010EA
60.5
EUR
ACRO360830010
ACRO360830050
2-Chlor-6-methyl-3-nitropyridin 98%
2-Chloro-6-methyl-3-nitropyridine
Formel:
C₆H₅ClN₂O₂ Molecular Weight: 172.57 g/mol Schmelzpunkt: 70…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03085820 CAS-Nummer: 56057-19-3 UN: 2811 ADR: 6.1 II |
Spezifikation Testergebnisse
Appearance | Yellow to brown Powder |
Infrared spectrum | Conforms |
Melting point | 69 to 74 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...