2-Chlor-6-methylbenzolsulfonylchlorid ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Chlor-6-methylbenzol-1-sulfonylchlorid, 2-Chlor-6-methylbenzolsulfochlorid, 2-Chlor-6-methylbenzolsulfonsäurechlorid
L07629.06
L07629.03
L07629.06EA
261
EUR
L07629.06
L07629.03
2-Chlor-6-methylbenzolsulfonylchlorid ≥98%
2-Chlor-6-methylbenzolsulfonylchlorid
Purity: 98%
CAS No.: 25300-37-2
Molecular Formula: C7H6Cl2O2S
Formula Weight: 225.09
Refractive Index: 1.5810
Boiling Point: 132-134°/0.5mm
UN No.: 3265
MDL No.: MFCD00051767
Formel:
C₇H₆Cl₂O₂S Molecular Weight: 225,09 g/mol Siedepunkt: 132…134 °C (0,5 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051767 CAS-Nummer: 25300-37-2 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...