2-Chlorbenzamidin Hydrochlorid
Lieferant: CHEMPUR
Synonyme:
2-Chlorbenzoesäureamidin Hydrochlorid, 2-Chlorphenylamidin Hydrochlorid
12-0016.1
12-0016.5
CHMP12-0016.1EA
0
EUR
CHMP12-0016.1
CHMP12-0016.5
2-Chlorbenzamidin Hydrochlorid
2-Chlorbenzamidin Hydrochlorid
Formel:
C₇H₈Cl₂N₂ Molecular Weight: 191,06 g/mol |
MDL:
MFCD00275607 CAS-Nummer: 18637-02-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...