2-Chlorimidazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chlor-1H-imidazol

M-Clarity™ Qualitätsstufe = MQ100

666408-1G 666408-250MG
SIAL666408-1GEA 155 EUR
SIAL666408-1G SIAL666408-250MG
2-Chlorimidazol, Sigma-Aldrich®
2-Chloroimidazole
Formel: C₃H₃ClN₂
Molecular Weight: 102,52 g/mol
MDL: MFCD02179530
CAS-Nummer: 16265-04-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR