2-Chlorthiobenzamid

Lieferant: FLUOROCHEM

Synonyme: 2-Chlorthiobenzoesäureamid

021317-1G 021317-2.5G
FLUO021317-1GEA 97.1 EUR
FLUO021317-1G FLUO021317-2.5G
2-Chlorthiobenzamid
2-Chlorthiobenzamid
Formel: C₇H₆ClNS
Molecular Weight: 171.65 g/mol
Siedepunkt: 316
Schmelzpunkt: 109…110 °C
MDL: MFCD00040955
CAS-Nummer: 15717-17-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR