2-Chlorthiobenzamid ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Chlorthiobenzoesäureamid
L11180.06
L11180.14
L11180.06EA
80.6
EUR
L11180.06
L11180.14
2-Chlorthiobenzamid ≥97%
2-Chlorthiobenzamid
Purity: 97%
CAS No.: 15717-17-6
Molecular Formula: C7H6ClNS
Formula Weight: 171.65
Melting Point: 63-66°
Beilstein Reference: 508511
Formel:
C₇H₆ClNS Molecular Weight: 171.65 g/mol Siedepunkt: 316 Schmelzpunkt: 63…66 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040955 CAS-Nummer: 15717-17-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...