5-Nitro-2-furonitril

Lieferant: Thermo Scientific

Synonyme: 2-Cyan-5-nitrofuran

RF02356.1 RF02356.10
MAYBRF02356.1EA 0 EUR
MAYBRF02356.1 MAYBRF02356.10
5-Nitro-2-furonitril
5-Nitro-2-furonitril
Formel: C₅H₂N₂O₃
Molecular Weight: 138,08 g/mol
Schmelzpunkt: 65 °C
MDL: MFCD00082647
CAS-Nummer: 59-82-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR