2-Furonitril ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Cyanfuran

A12214.06 A12214.14
A12214.06EA 76.5 EUR
A12214.06 A12214.14
2-Furonitril ≥98%
2-Furonitril

Purity: 98+% CAS No.: 617-90-3 Molecular Formula: C5H3NO Formula Weight: 93.09 Flash Point: 35°(95°F) Refractive Index: 1.4790 Odour: Recognizable Boiling Point: 146-148° Beilstein Reference: 107033 UN No.: 1992 MDL No.: MFCD00003223

Formel: C₅H₃NO
Molecular Weight: 93,08 g/mol
Siedepunkt: 146…148 °C
Dichte: 1,062
Flammpunkt: 35 °C (95 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003223
CAS-Nummer: 617-90-3
EINECS: 210-537-4
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR