2-Cyanphenylboronsäure
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
Dihydroxy(2-cyanphenyl)boran, 2-Cyanbenzolboronsäure, (2-Cyanphenyl)metaborsäure
BB-2471-5G
BB-2471-25G
BB-2471-1G
COBBBB-2471-5GEA
154
EUR
COBBBB-2471-5G
COBBBB-2471-25G
COBBBB-2471-1G
2-Cyanphenylboronsäure
2-Cyanphenylboronsäure
Formel:
C₇H₆BNO₂ Molecular Weight: 146,94 g/mol |
MDL:
MFCD01632208 CAS-Nummer: 138642-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...