2-Acetyl-5-methylfuran, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(5-Methylfuran-2-yl)ethanone

M-Clarity™ Qualitätsstufe = MQ100

299553-25G 299553-5G
SIAL299553-25GEA 56.8 EUR
SIAL299553-25G SIAL299553-5G
2-Acetyl-5-methylfuran, Sigma-Aldrich®
2-Acetyl-5-methylfuran
Formel: C₇H₈O₂
Molecular Weight: 124,14 g/mol
MDL: MFCD00003243
CAS-Nummer: 1193-79-9
EINECS: 214-779-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR