2-Acetylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Acetylbenzolboronsäure, Dihydroxy(2-acetylphenyl)boran, (2-Acetylphenyl)metaborsäure

BB-2185-25G BB-2185-1G BB-2185-5G
COBBBB-2185-25GEA 2040 EUR
COBBBB-2185-25G COBBBB-2185-1G COBBBB-2185-5G
2-Acetylphenylboronsäure
2-Acetylphenylboronsäure
Formel: C₈H₉BO₃
Molecular Weight: 163,97 g/mol
MDL: MFCD01321263
CAS-Nummer: 308103-40-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR