2-Vinylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Dihydroxy(2-vinylphenyl)borane, (2-Vinylphenyl)metaboric acid, 2-Vinylbenzeneboronic acid

BB-2200-1G BB-2200-5G
COBBBB-2200-1GEA 153 EUR
COBBBB-2200-1G COBBBB-2200-5G
2-Vinylphenylboronsäure
2-Vinylphenylboronsäure
Formel: C₈H₉BO₂
Molecular Weight: 147,97 g/mol
MDL: MFCD01074659
CAS-Nummer: 15016-42-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR