2-Ethoxy-5-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Dihydroxy(2-ethoxy-5-fluorphenyl)boran, 2-Ethoxy-5-fluorbenzolboronsäure, (2-Ethoxy-5-fluorphenyl)metaborsäure

BB-8050-5G BB-8050-1G BB-8050-25G
COBBBB-8050-5GEA 93.7 EUR
COBBBB-8050-5G COBBBB-8050-1G COBBBB-8050-25G
2-Ethoxy-5-fluorphenylboronsäure
2-Ethoxy-5-fluorphenylboronsäure
Formel: C₈H₁₀BFO₃
Molecular Weight: 183.97 g/mol
MDL: MFCD05664328
CAS-Nummer: 864301-27-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR