2-Ethoxy-6-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3954-5G BB-3954-25G BB-3954-1G
COBBBB-3954-5GEA 61.9 EUR
COBBBB-3954-5G COBBBB-3954-25G COBBBB-3954-1G
2-Ethoxy-6-fluorphenylboronsäure
2-Ethoxy-6-fluorphenylboronsäure
Formel: C₈H₁₀BFO₃
MDL: MFCD05664322
CAS-Nummer: 957062-68-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR