1,1,1-Tris(hydroxymethyl)propan ≥98%

Lieferant: Thermo Scientific

Synonyme: Propylidintrimethanol, Trimethylolpropan, 2-Ethyl-2-(hydroxymethyl)-1,3-propandiol

A13084.0C A13084.0E A13084.36
A13084.0CEA 343 EUR
A13084.0C A13084.0E A13084.36
1,1,1-Tris(hydroxymethyl)propan ≥98%
1,1,1-Tris(hydroxymethyl)propan

Purity: 98% CAS No.: 77-99-6 Molecular Formula: C6H14O3 Formula Weight: 134.18 Flash Point: 172°(341°F) Melting Point: 59-60° Boiling Point: 296-298° Beilstein Reference: 1698309 MDL No.: MFCD00004694

Formel: C₆H₁₄O₃
Molecular Weight: 134,18 g/mol
Siedepunkt: 296…298 °C
Schmelzpunkt: 59…60 °C
Dichte: 1,15 g/cm³ (20 °C)
Flammpunkt: 172 °C (341 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004694
CAS-Nummer: 77-99-6
EINECS: 201-074-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR