2-Ethylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Ethylphenyl)metaboric acid, 2-Ethylbenzeneboronic acid, Dihydroxy(2-ethylphenyl)borane

BB-2611-5G BB-2611-1G
COBBBB-2611-5GEA 59.3 EUR
COBBBB-2611-5G COBBBB-2611-1G
2-Ethylphenylboronsäure
2-Ethylphenylboronsäure
Formel: C₈H₁₁BO₂
Molecular Weight: 149.98 g/mol
MDL: MFCD02093075
CAS-Nummer: 90002-36-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR