2-Fluor-3-(trifluormethyl)pyridin
Lieferant: FLUOROCHEM
013923-25G
013923-5G
013923-1G
FLUO013923-25GEA
221
EUR
FLUO013923-25G
FLUO013923-5G
FLUO013923-1G
2-Fluor-3-(trifluormethyl)pyridin
2-Fluoro-3-(trifluoromethyl)pyridine
Formel:
C₆H₃F₄N Molecular Weight: 165.09 g/mol |
MDL:
MFCD04973416 CAS-Nummer: 65753-52-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...