2-Fluor-m-anisaldehyd 98%

Lieferant: COMBI-BLOCKS
Gefahr

Synonyme: 2-Fluor-3-methoxybenzaldehyd, 2-Fluor-3-anisaldehyd

OT-0035-5G OT-0035-25G OT-0035-1G
COBBOT-0035-5GEA 127 EUR
COBBOT-0035-5G COBBOT-0035-25G COBBOT-0035-1G
2-Fluor-m-anisaldehyd 98%
2-Fluor-m-anisaldehyd
Formel: C₈H₇FO₂
Molecular Weight: 154.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03411962
CAS-Nummer: 103438-88-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR