2-Fluor-3-methylpyridin

Lieferant: COMBI-BLOCKS
Gefahr

Synonyme: 2-Fluor-3-picolin

OS-7441-5G OS-7441-25G OS-7441-100G
COBBOS-7441-5GEA 25.5 EUR
COBBOS-7441-5G COBBOS-7441-25G COBBOS-7441-100G
2-Fluor-3-methylpyridin
2-Fluor-3-methylpyridin
Formel: C₆H₆FN
Molecular Weight: 111,12 g/mol
MDL: MFCD00041223
CAS-Nummer: 2369-18-8
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR