2-Fluor-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluor-4-pyridinylboronsäure, 2-Fluorpyridin-4-boronsäure

BB-2916-5G BB-2916-1G
COBBBB-2916-5GEA 181 EUR
COBBBB-2916-5G COBBBB-2916-1G
2-Fluor-4-pyridinboronsäure
2-Fluoro-4-pyridineboronic acid
Formel: C₅H₅BFNO₂
Molecular Weight: 140,91 g/mol
MDL: MFCD04112534
CAS-Nummer: 401815-98-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR