2-Fluor-5-(trifluormethyl)pyridin

Lieferant: COMBI-BLOCKS

OS-7604-5G OS-7604-25G OS-7604-1G
COBBOS-7604-5GEA 35.9 EUR
COBBOS-7604-5G COBBOS-7604-25G COBBOS-7604-1G
2-Fluor-5-(trifluormethyl)pyridin
2-Fluoro-5-(trifluoromethyl)pyridine
Formel: C₆H₃F₄N
Molecular Weight: 165.09 g/mol
MDL: MFCD07437945
CAS-Nummer: 69045-82-5
EINECS: 400-290-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR