2-Fluor-5-hydroxypyridin
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
2-Fluor-5-pyridinol
PY-7275-5G
PY-7275-1G
COBBPY-7275-5GEA
97
EUR
COBBPY-7275-5G
COBBPY-7275-1G
2-Fluor-5-hydroxypyridin
2-Fluor-5-hydroxypyridin
Formel:
C₅H₄FNO Molecular Weight: 113.09 g/mol |
MDL:
MFCD01075040 CAS-Nummer: 55758-32-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...