2-Fluor-5-iodbenzaldehyd ≥97%

Lieferant: Thermo Scientific
Achtung

H32033.06 H32033.14
H32033.06EA 91.7 EUR
H32033.06 H32033.14
2-Fluor-5-iodbenzaldehyd ≥97%
2-Fluoro-5-iodobenzaldehyde

Purity: 97+% CAS No.: 146137-76-0 Molecular Formula: C7H4FlO Formula Weight: 250.01 Melting Point: 42-46° MDL No.: MFCD03093999

Formel: C₇H₄FIO
Molecular Weight: 250,01 g/mol
Schmelzpunkt: 42…46 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03093999
CAS-Nummer: 146137-76-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR