2-Fluor-5-nitrobenzoesäure

Lieferant: COMBI-BLOCKS
Achtung

AN-1060-100G AN-1060-5G AN-1060-25G
COBBAN-1060-100GEA 196 EUR
COBBAN-1060-100G COBBAN-1060-5G COBBAN-1060-25G
2-Fluor-5-nitrobenzoesäure
2-Fluor-5-nitrobenzoesäure
Formel: C₇H₄FNO₄
Molecular Weight: 185,11 g/mol
MDL: MFCD00134238
CAS-Nummer: 7304-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR