2-Fluor-6-methylpyridin

Lieferant: COMBI-BLOCKS
Gefahr

Synonyme: 2-Fluor-6-picolin

OS-7545-100G OS-7545-25G
COBBOS-7545-100GEA 169 EUR
COBBOS-7545-100G COBBOS-7545-25G
2-Fluor-6-methylpyridin
2-Fluor-6-methylpyridin
Formel: C₆H₆FN
Molecular Weight: 111.12 g/mol
MDL: MFCD00041226
CAS-Nummer: 407-22-7
EINECS: 206-980-8
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR