2-Fluorbenzamidin Hydrochlorid
Lieferant: CHEMPUR
Synonyme:
2-Fluorbenzoesäureamidin Hydrochlorid, 2-Fluorphenylamidin Hydrochlorid
12-0051.1
12-0051.5
CHMP12-0051.1EA
0
EUR
CHMP12-0051.1
CHMP12-0051.5
2-Fluorbenzamidin Hydrochlorid
2-Fluorbenzamidin Hydrochlorid
Formel:
C₇H₈ClFN₂ Molecular Weight: 174,61 g/mol |
MDL:
MFCD04114435 CAS-Nummer: 57075-81-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...