2-Fluorbenzolsulfonamid

Lieferant: Thermo Scientific

KM09947.10 KM09947.50
MAYBKM09947.10EA 0 EUR
MAYBKM09947.10 MAYBKM09947.50
2-Fluorbenzolsulfonamid
2-Fluorbenzolsulfonamid
Formel: C₆H₆FNO₂S
MDL: MFCD00042283
CAS-Nummer: 30058-40-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR