2-Fluorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2-Fluorphenyl)metaborsäure, 2-Fluorbenzolboronsäure, Dihydroxy(2-fluorphenyl)boran
BB-2658-5G
BB-2658-25G
BB-2658-100G
COBBBB-2658-5GEA
26.2
EUR
COBBBB-2658-5G
COBBBB-2658-25G
COBBBB-2658-100G
2-Fluorphenylboronsäure
2-Fluorphenylboronsäure
Formel:
C₆H₆BFO₂ Molecular Weight: 139,92 g/mol |
MDL:
MFCD00674013 CAS-Nummer: 1993-03-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...