Furan-2-boronsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Furan-2-ylboronic acid
359390010
359390100
ACRO359390010EA
52.3
EUR
ACRO359390010
ACRO359390100
Furan-2-boronsäure 97%
Furan-2-boronsäure
Formel:
C₄H₅BO₃ Molecular Weight: 111,89 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00799544 CAS-Nummer: 13331-23-2 |
Spezifikation Testergebnisse
Appearance | Light beige Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...