Furan-2-boronsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Furan-2-ylboronic acid

B23842.03 B23842.06 B23842.14
B23842.03EA 54 EUR
B23842.03 B23842.06 B23842.14
Furan-2-boronsäure ≥97%
Furan-2-boronsäure

Purity: 97% CAS No.: 13331-23-2 Molecular Formula: C4H5BO3 Formula Weight: 111.89 Storage Temperature: -30°C to -10°C Melting Point: ca 120° dec. Beilstein Reference: 112370 MDL No.: MFCD00799544

Formel: C₄H₅BO₃
Molecular Weight: 111.89 g/mol
Schmelzpunkt: ∼ 120 °C (Zers.)
MDL: MFCD00799544
CAS-Nummer: 13331-23-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR